Learn More
Fisherbrand™ accuflex™ Electrode Support
For accumet™ benchtop meters
Brand: Fisherbrand™ 01X321801
This item is not returnable.
View return policy
Description
For accumet™ benchtop meters
- Attaches to meter base with bracket supplied
- Holds one micro electrode and three standard electrodes
- Permits full up/down and left/right positioning of electrodes
- Made of chemical-resistant plastic
Specifications
Support Arm and Bracket | |
pH | |
Electrode support arm bracket for meter | |
Plastic |
Fisherbrand Focus on pH and electrochemistry
File size: 9.1 MB
Fisherbrand Focus on pH and electrochemistry
File size: 2.5 MB
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
Product Content Correction
Product Title
Fisherbrand™ accuflex™ Electrode Support > Accuflex Electrode Support Arm
Spot an opportunity for improvement?Share a Content Correction