Learn More
Fisherbrand™ Maxima C Plus Centering Rings
Centering Ring for use with Maxima C vacuum pumps to join two flanges
Brand: Fisherbrand™ FB70127
This item is not returnable.
View return policy
Description
For use with Model M2C and M4C vacuum pump
Specifications
Model M2C and M4C vacuum pump | |
Stainless Steel | |
Centering Ring |
Product Content Correction
Product Title
Fisherbrand™ Maxima C Plus Centering Rings > Size: NW 16; Fit Models: M2C and M4C
Spot an opportunity for improvement?Share a Content Correction