Learn More
Fisherbrand™ Maxima C Plus Centering Rings
Centering Ring for use with Maxima C vacuum pumps to join two flanges
Brand:  Fisherbrand™ 701073
75.36 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10534002
  • 77.30 DKK / Each
Only null left
Estimated Shipment: 12-08-2025
Description
For use with Model M2C and M4C vacuum pump
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Model M8C, M12C and M16C vacuum pump
Stainless Steel
Centering Ring
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Maxima C Plus Centering Rings > Size: NW 25; Fit Models: M8C, M12C, and M16C