Learn More
Fisherbrand™ MultiTube Vortexers: Post Extension Kit
Post extension kit for Fisherbrand™ MultiTube Vortexers.
Brand: Fisherbrand™ 80862908
1188.37 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
For use with Fisher Scientific MultiTube Vortexers
- Post extension kit adds 6 in. (15.2cm) to post to accommodate tubes up to 25.4cm (10 in.)
Specifications
Post Extension Kit | |
Multitube vortexer |
Product Content Correction
Product Title
Fisherbrand™ MultiTube Vortexers: Post Extension Kit > Post Extension Kit
Spot an opportunity for improvement?Share a Content Correction