missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Aluminum Foil, Standard-Gauge Roll

For sealing, weighing, and transferring samples

Brand:  Fisherbrand™ 01213105

 View more versions of this product

Product Code. 15397768

  • 555.00 DKK / Each
Only null left
Estimated Shipment: 14-08-2025
Add to basket
This item is not returnable. View return policy

Description

Description

Ideal for weighing, sealing, and tranferring your samples.

  • Packed in individual dispenser box with cutter bar
  • Heavy-gauge rolls offer greater folding strength, good for covering sharp edges
Specifications

Specifications

Sealing, Weighing, Transferring Samples
Foil
46cm
Aluminum
492 ft.
18 in.
149.9 m
24 ÎĽm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Aluminum Foil, Standard-Gauge Roll > Heavy-gauge roll; Width: 45.7cm; Length: 152.4m

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.