Learn More
Fisherbrand™ Lens Paper Product Code.: 10216202
Estimated Shipment: 22-04-2025
to see stock.
Add to basket
Fisherbrand™ Lens Paper
Product Code. 10216202
4 x 6 in., 50 Sheets/Book, 12 Books/Pk.
Click to view available options
Dimensions (L x W):
4 x 6 in.
6 x 8 in. sheets
Quantity:
100 Sheets/Book, 12 Books/Pk.
50 Sheets/Book, 12 Books/Pk.
Product Code. 10216202
Estimated Shipment: 22-04-2025
to see stock.
Add to basket
Add to basket
For cleaning glass lenses
Booklet
  • Never scratches glass lenses, objectives, or other delicate surfaces
  • Doesn't become harsh; doesn't readily collect dust or grease
  • Virtually lint-free sheets are glued and bound in a booklet for easy removal
Dispenser Box
  • Same paper as above, but packaged in a convenient dispenser box for fast, single-sheet removal individual sheets measure 5L × 7 in.W (13 × 18cm)
Specifications
Product Type Lens Cleaning Paper
Dimensions (L x W) 4 x 6 in.
For Use With (Application) For cleaning Glass lenses
Quantity 50 Sheets/Book, 12 Books/Pk.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number9296216 Certificate TypeCertificate of Analysis Date08/06/2021 Catalog Number11996
Product Content Correction
Product Title
Fisherbrand™ Lens Paper >