Learn More
Fisherbrand™ Plastic Joint Clips
Made of plastic for use on standard taper glass joints
117.00 DKK
Product Code | Brand | Color | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | For Use With (Equipment) | Price | Quantity & Availability | |||||
17555156
|
Fisherbrand™
14-955-256 |
Red | Standard taper Glass joint size 29 |
117.00 DKK
Pack of 10 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
|||||
Description
Made of plastic for use on standard taper glass joints
- Color coded for easy size identification
- Does not corrode
- Will not scratch glass
- Withstands temperatures up to 150°C
Specifications
10/Pk. | |
Joint Clip | |
Plastic |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Plastic Joint Clips