Learn More
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Strong and leak-resistant
Brand: Fisherbrand™ 22044563
800.85 DKK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
Description
Strong and leak-resistant
- Printed with biohazard symbol and text in English and Spanish
- Supplied with twist tie closure
Specifications
Twist Tie | |
23 x 19 in. (58 x 48 cm) | |
48 cm | |
Biohazard symbol and text in English and Spanish | |
23 in. | |
Polyethylene |
Red with Black Print | |
58 cm | |
Autoclavable | |
19 in. | |
1.25 mil |
Promotions
-
Multiple Manufacturers
Everyday Lab Essentials
From wipes to waste bins, labels to liners – save on them all.Promo Code: 24132Expires: 2025-08-24
New Offer Shop Now