Learn More
Fisherbrand™ Covers for Fisherbrand™ Ultrasonic Cleaners
A practical cover to complete your S-Line Ultrasonic Cleaning Unit. Fisherbrand™ Covers for S-Line Ultrasonic Cleaning Units are manufactured from durable blue plastic.
Brand:  Fisherbrand™ 1009050
 View more versions of this product
Product Code. 11706208
  • 574.00 DKK / Each
Only null left
Estimated Shipment: 08-07-2025
Add to basket
Description
Description
  • Easy to clean
  • Clean-cut, smooth surfaces
  • Meets high user demands for hygiene
Specifications
Specifications
Lid
Ultrasonic Cleaner, 45 L
Stainless Steel
Videos
  • Fisherbrand - Ultrasound for laboratory applications and for intensive cleaning
    Fisherbrand - Ultrasound for laboratory applications and for intensive cleaning
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Fisherbrand Ultrasound for laboratory applications and for intensive cleaning
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Covers for Fisherbrand™ Ultrasonic Cleaners > For S 450 H Ultrasonic Baths