Learn More
Mettler Toledo™ Sample Bottles for FiveGo™ pH/Conductivity/DO Meter
For use with FiveGo pH/Conductivity/DO Meters.
Brand: Mettler Toledo™ 30239143
171.60 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Quantity: 4/Pk.
Specifications
Sample Bottle |
Promotions
-
Multiple Manufacturers
Focus On Clean Water
Shop now for up to 60% off Clean Water Research essentialsPromo Code: 24139Expires: 2025-08-21
Days
33Hours
9Minutes
58Seconds
11New Offer Save Now