Learn More
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments are accurate and reproducible over a range of volumes.
Brand:  Thermo Scientific™ 36520050
1392.21 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10350925
  • 1400.00 DKK / Each
Description
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments are accurate and reproducible over a range of volumes.
  • Manual syringes in industry-standard size
  • White scale backing for easy reading
  • Solvent-resistant markings
TRUSTED_SUSTAINABILITY
Specifications
Specifications
36520050
50 mm
Reusable
100 ÎĽL
Side Hole
Removable
23 G
Gas Tight GC Autosampler Syringe, Removable Needle
Ea.
50 mm
TriPlus, AS200/AS800, AI/AS 1610 Autosampler
36550040
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments > 100ÎĽL; 23 gauge; 50mm