Learn More
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments are accurate and reproducible over a range of volumes.
Brand: Thermo Scientific™ 36500495
963.21 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments are accurate and reproducible over a range of volumes.
- Manual syringes in industry-standard size
- White scale backing for easy reading
- Solvent-resistant markings
Specifications
36500495 | |
Gas Tight GC Autosampler Syringe, Removable Needle | |
Ea. | |
50 mm | |
TriPlus, AS2000, AS200/AS800 Autosampler | |
36566485 |
23 G | |
Reusable | |
100 ÎĽL | |
Cone | |
Removable |