Learn More
Fisherbrand™ Fixed Tube Rack
For use with Fisherbrand™ Multi-Platform Shaker and Microplate Mini Shaker
Brand:  Fisherbrand™ 88861207
588.05 DKK valid until 2025-01-31
Use promo code "24160" to get your promotional price.
 View more versions of this product
Product Code. 15793798
  • 619.00 DKK / Each
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Maximum 2 per platform
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fixed Tube Rack
27
262 x 112 x 140 mm
27 x 25 mL Tubes, Tubes (25 mL)
Fisherbrand™ Multi-Platform Shaker and Microplate Mini Shaker
22 mm