Learn More
Thermo Scientific™ Flat PCR Caps, strips of 8, Sustain™ Series
Thermo Scientific Flat PCR Caps, strips of 8, Sustain Series, are designed to work with PCR plastics and Sustain Series standard low profile strip tubes and 96-well plates.
322.00 DKK - 696.00 DKK
Specifications
Color | Natural |
---|---|
Certifications/Compliance | Certified DNase-, RNase-, Human DNA-, and PCR Inhibitor-free |
Material | Polypropylene |
Autoclavable | Autoclavable |
Green Features | Less waste, renewable material |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
30179689
![]() |
Thermo Scientific™
AB0783SS |
120 strips |
322.00 DKK
Pack of 120 |
In Stock
Log in to see stock. |
||||||
30179685
![]() |
Thermo Scientific™
AB0784SS |
250 strips |
696.00 DKK
Pack of 250 |
In Stock
Log in to see stock. |
||||||
Description
Thermo Scientific Flat PCR Caps, strips of 8, Sustain Series, are designed to work with PCR plastics and Sustain Series standard low profile strip tubes and 96-well plates. The strip caps are easy to apply and create a secure seal to prevent minimize evaporation.
Features and benefits
- Suitable for PCR applications
- Flat surface of each cap facilitates sample labeling
- Certified DNase-, RNase-, human DNA-, and PCR inhibitor-free
A breakthrough in sustainable PCR technology
Thermo Scientific Sustain Series PCR plastics signify a breakthrough in our PCR and quantitative PCR (qPCR) product offering that goes beyond providing accurate and reproducible PCR data. They represent our commitment to integrating more sustainable practices into our manufacturing processes, without compromising on product quality or performance. Our Sustain Series PCR plastics are manufactured under ISO 9001 guidelines within a Class 100,000 cleanroom from a biobased medical-grade virgin polypropylene that is chemically and molecularly identical to our existing Thermo Scientific PCR plastics. These products are supported by certification from the globally recognized International Sustainability and Carbon Certification (ISCC) system. This greener approach is equivalent to 1.96X lower CO2 equivalent emissions per kg of biobased polypropylene resin compared to traditional fossil fuel-based polypropylene resin.
Additional product details
- Uniform, ultra-thin walls enable maximum and consistent heat transfer for equally high performance of every sample
- Each well is visually inspected and electrostatically tested for defects and contaminants, and every lot is run through a PCR cycling test to evaluate sealing
- Manufactured from second generation bio-circular feedstocks that are chemically and molecularly identical to our existing Thermo Scientific PCR plastics
Specifications
Natural | |
Polypropylene | |
Less waste, renewable material |
Certified DNase-, RNase-, Human DNA-, and PCR Inhibitor-free | |
Autoclavable |
For Laboratory Use