Learn More
Memmert™ Flexible Pt100 Temperature Sensor for UF1060
Flexible temperature sensor for positioning in chamber or in load.
Brand: Memmert™ UF1060H8
This item is not returnable.
View return policy
Description
- Positioned flexibly in chamber or load
- For local temperature measurement (up to 1 additional sensor is possible)
- The measured temperature will be indicated on the display, recorded in the integral data store, and can be documented via the AtmoCONTROL software
- For Memmert Ovens
Specifications
UF1060 Oven | |
Temperature Sensor |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Flexible Pt100 Temperature Sensor for UF1060 > For Use With: UF1060
Spot an opportunity for improvement?Share a Content Correction