Learn More
FLI1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11347-1-AP-20UL
 View more versions of this product
Product Code. 16828363
  • 1081.84 DKK / 20µL
This item is not returnable. View return policy
Description
Description
ETS-1 is the prototype member of a family of genes identified on the basis of homology to the v-Ets oncogene isolated from the E26 erythroblastosis virus. This family of genes currently includes ETS-1, ETS-2, ERG-1, ERG-2, ELK, E74, FLI-1, PU. 1 and PEA3. Members of the ETS gene family exhibit varied pattern of tissue expression, and share a highly conserved carboxy terminal domain containing a sequence related to the SV40 large T antigen nuclear localization signal sequence.
Specifications
Specifications
FLI1
Polyclonal
Unconjugated
FLI1
EWSR2, FLI1, Proto oncogene Fli 1, SIC 1, Transcription factor ERGB
Rabbit
Antigen Affinity Chromatography
RUO
14247, 2313
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.53 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P26323, Q01543
FLI1
FLI1 Fusion Protein Ag1899
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FLI1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated