Learn More
Scotch-Brite™ Floor Pads, Green
For the efficient, intermediate cleaning of heavily soiled floors with or without a coating. For use with single disc machines or automatic floor cleaning machines with slow speed.
Brand:  Scotch-Brite™ 7100159881
755.25 DKK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
 View more versions of this product
Product Code. 17118810
  • 795.00 DKK / Pack of 5
Everyday Lab Essentials
Use promo code 24132 at checkout
View Available Promotions
Only null left
Estimated Shipment: 05-09-2025
Specifications
Floor Pad
432 mm
Single Disc Machines or Slow Speed Automatic Floor Cleaning Machines
Green
Scrubbing
Round
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    From wipes to waste bins, labels to liners – save on them all.
    Promo Code: 24132
    Expires2025-08-24
    New Offer
    Shop Now
Product Content Correction
Product Title
Scotch-Brite™ Floor Pads, Green > 432 mm