Learn More
WTW™ Flow-through Vessel for Conductivity Cells
Flow vessels for WTW conductivity measuring cells are perfectly designed to perform measurements without the interference of carbon dioxide through the air or to examine the conductivity of solutions in closed circuits.The overpressure in the system must not exceed 0.5 bar.
Brand:  WTW™ 302750
1581.72 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10671832
  • 1660.00 DKK / Each
In Stock
Explore more special offers
Specifications
Flow-through vessel
18 mm
Product Title
WTW™ Flow-through Vessel for Conductivity Cells > Diameter: 18mm