Learn More
Mettler Toledo™ Fluoroborate Half Cell Electrode
Brand:  Mettler Toledo™ 51107676
10072.30 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11947701
  • 10330.00 DKK / Each
Only null left
Estimated Shipment: 21-07-2025
Specifications
Polymer Membrane
0°C to 50°C
2 to 12
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Mettler Toledo™ Fluoroborate Half Cell Electrode > Membrane: Polymer