Learn More
Memmert™ Flush-Fit Set (Frame Covering Gap Between Oven And Wall Opening, W/Air Slots) For S450
Brand: Memmert™ S450B29740
This item is not returnable.
View return policy
Specifications
Models U, UF TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 450 Series Model | |
Flush-Fit Unit | |
Stainless Steel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Flush-Fit Set (Frame Covering Gap Between Oven And Wall Opening, W/Air Slots) For S450 >
Spot an opportunity for improvement?Share a Content Correction