Learn More
Sartorius™ Forceps with Silicone Covered Tips
For use with balance weights
Brand:  Sartorius™ YAW31
193.98 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11397504
  • 199.00 DKK / Each
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Description
For use with balance weights
  • Prevents damage to the surface of weights
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Each
105 mm
Forceps with Silicone
Forceps with Silicone
Product Title
Sartorius™ Forceps with Silicone Covered Tips > Length: 105mm