Learn More
Cole-Parmer™ Forceps, Reinforced Nylon
Disposable nylon forceps with serrated, round tips. Cole-Parmer™ Forceps, Reinforced Nylon feature corrugated handles and serrated blades and rounded tips for handling fragile samples.
Brand:  Cole-Parmer™ 06443-15
Product Code. 12895043
  • 1220.00 DKK / Pack of 100
Only null left
Estimated Shipment: 17-09-2025
Description
  • White nylon construction
  • Corrugated handles
  • Serrated blades
  • Autoclavable
Specifications
Specifications
Round
Corrugated and Serrated
13.38 cm
100/Pack
Useful in Handling Fragile Samples
Serrated
100
Nylon
Forceps
Corrugated and Serrated
Forceps
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Cole-Parmer™ Forceps, Reinforced Nylon > Length: 4.9mm