Learn More
Foss™ Digestion System: Digestion Tubes
Straight-sided tubes recommended for the majority of digestion applications
Brand: Foss™ TC1000-0735
This item is not returnable.
View return policy
Description
- Volumetric tubes with constriction at neck recommended for applications where the digestate requires dilution to a fixed volume before analyses such as FIA, SFA, and AA
Specifications
Straight-sided Tube | |
100 mL | |
40/Pk. |
3.38 oz. | |
Digestion applications |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.