Learn More
Cytiva Frac30 Model Fraction Collector
Used as a round fraction collector. Cytiva Frac30 Model Fraction Collector allows you to collect up to 30 fractions.
Brand: Cytiva 29023051
12466.35 DKK valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Description
- Controlled via the AKTA protein purification system start touch screen instrument display
- Also controlled by using UNICORN start software via computer
- Collect up to 30 fractions
- Supports four tube sizes, ranging from 1.5mL to 15mL
- Fractions can be collected in volumes ranging from 0.5mL to 15mL
- The entire fraction collector rack can be transferred as required
Specifications
Fraction Collector |
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
26Hours
4Minutes
28Seconds
58