Learn More
Thermo Scientific™ Frames and Caps for Immuno Standard Modules
Easily insert and remove strip plates with solid modules by utilizing these frames and caps for immuno standard modules.
Brand: Thermo Scientific™ 430805
1126.61 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Easily insert and remove strip plates with solid modules by utilizing Thermo Scientific™ Frames and Caps for Immuno Standard Modules.
- Polyethylene, stackable frame is marked for lid orientation
Specifications
Strip Plate | |
2 Pk. | |
Strip Cap | |
8 | |
120 | |
Immuno Standard Modules |
1 x 8 | |
Clear | |
Polyethylene | |
Sterile | |
2 | |
120/Cs. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1319944 | Certificate of Processing | 04/12/2024 | 430805 |
1311023 | Certificate of Processing | 04/12/2024 | 430805 |
1319944 | Certificate of Compliance | 03/06/2021 | 430805 |
1311023 | Certificate of Compliance | 23/02/2021 | 430805 |
1262906 | Certificate of Compliance | 21/08/2019 | 430805 |