Learn More
BUCHI Freeze Drying Beaker Flasks and Accessories
Freeze drying beaker flasks and accessories for use with a lyophilizer.
Brand:  BUCHI 11065730
 View more versions of this product
Product Code. 17254664
  • 625.00 DKK / Each
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Description
  • For use with a lyophilizer
Specifications
Specifications
800 to 1200 mL
L-200
110 mm
0.7 lb.
Freeze Drying
Beaker Flask Cap
-60°C to +30°C
0.4 kg