Learn More
Eppendorf™ Freezer Rack: Innova™ C580, C760, 2 in/53 mm
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Brand:  Eppendorf™ 6001040211
Product Code. 15753255
  • 1089.34 DKK / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Freezer Rack: Innova™ C580, C760, locking rod, 2 in/53 mm, side access, stainless steel- Rack for 53 mm/2 in boxes- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 148 x 141 x 776 (5.8 x 5.6 x 30.6)- Max. box number per rack: 14 (14 x 1)- Stainless steel- For ULT freezer: Innova C580 (max. 32 racks), Innova C760 (max. 40 racks)- For ULT freezer: Innova C580 (max. 448 boxes), Innova C760 (max. 560 boxes)- With locking rod for increased sample safety
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Freezer Rack