Learn More
IKA™ Fritte Set
This Fritte Set contains two glass frits for the Pro-Divide reaction cell.
Brand:  IKA™ 0020102633
 View more versions of this product
Product Code. 17573626
  • 535.00 DKK / Pack of 2
In Stock
Explore more special offers
Description
  • This set contains two glass frits for the Pro-Divide reaction cell.
  • The size of each pore falls between these ranges:
    • 1 μm set: 1 and 1.6 μm
    • 5 μm set: 4 and 5.5 μm
    • 10 μm set: 10 and 16 μm
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fritte Set
5 μm
Pro-Divide Reaction Cell
Documents
One moment while we fetch your results.
Product Title
IKA™ Fritte Set > Fritte Set; Size: 5 μm