Learn More
CleanSpace Technology™ Full Face Mask Quantitative Fit Test PortaCount Adaptor Product Code.: 15950082
Only undefined left
Add to basket
CleanSpace Technology™ Full Face Mask Quantitative Fit Test PortaCount Adaptor
Product Code. 15950082 Shop All CleanSpace Technology Products
For use with: CleanSpace Full Face Mask (PAF-1014)
Click to view available options
:
For use with: CleanSpace Full Face Mask (PAF-1014)
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15950082
Brand: CleanSpace Technology™ PAF1015
Only undefined left
Add to basket
Only undefined left
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
The CleanSpace Full Face Mask Quantitative Fit Test (PortaCount) Adaptor is designed to be used with the CleanSpace Full Face Mask (PAF-1014). It is utilised as a mask fit test adaptor to be fitted in the front of the mask for Quantitative Fit (TSI PortaCount) Testing.
  • Used with the revolutionary CleanSpace PAPR: light weight, no hoses or belts
  • Designed for comfort over long periods
  • Allows sweating and breathability
  • Easy to wash and quick drying
  • Designed for long wear in harsh environments
  • Easy and quick replacement
Specifications
For Use With (Equipment) Obsolete
Product Content Correction
Product Title
CleanSpace Technology™ Full Face Mask Quantitative Fit Test PortaCount Adaptor >