Learn More
3M™ Full Facepiece Reusable Respirator
3M™ Promask™ Full Facepiece Reusable Respirator FM3 is a multi-function full-face mask designed to provide durable and comfortable protection against a wide range of respiratory hazards. Use in conjunction with approved 3M powered air systems or as a negative pressure respirator.
Brand: 3M™ 7100236989
4385.33 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
View more versions of this product
Description
- Use as a negative-pressure single-filter mask or as a face piece for a powered air respirator
- Sweat port helps to provide user comfort during long periods of use
- Visor provides a good field of view and comes in choice of polycarbonate or high-heat material
- Side-fitting filter port and clear inner mask for good forwards and downwards vision
- Filter doesn't interfere with front-mounted speech diaphragm to aid in easy communication
Specifications
Bayonet & DIN | |
Full Face Mask | |
FF-302 | |
5 Point Strap |
Black | |
Large | |
Yes |