missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUS/TLS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11570-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is a rabbit polyclonal antibody raised against an internal region of human FUS.
FUS/TLS was originally described as fused in Sarcoma is actually related to a number of different cellular processes and correlated to several diseases. FUS binds DNA as a single or double strand, and acts as a site for transcription factors to recognize. Since FUS is well-documented to be involved in liposarcoma, lipoma and leukaemia, it is believed that when its normal housekeeping functions related to transcription are disrupted, normal cells become cancerous. Interestingly, FUS is also associated with the fatal neuron disorder amyotrophic lateral sclerosis; interruption of its normal RNA-binding functions leads to incorrect cellular localization and leads to pathologies of the neuron. Thus, FUS regulation of cellular functions is a critical component of maintece of healthy cells.Specifications
FUS/TLS | |
Polyclonal | |
Unconjugated | |
FUS | |
75 kDa DNA pairing protein, FUS, FUS1, FUS-CHOP, hnRNP P2, Oncogene FUS, Oncogene TLS, POMp75, RNA binding protein FUS, TLS, TLS/CHOP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
233908, 2521, 317385 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P35637, P56959 | |
Fus | |
FUS/TLS Fusion Protein Ag2150 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FUS/TLS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction