Learn More
A&D Weighing FX/FZ Precision Balances Class II Approved
GREENER_CHOICE
A&D Weighing FZ/FX Series Precision Balances combine all of the essential weighing functions in a compact, light-weight package.
Brand:  A&D Weighing FZ-3000i-EC
 View more versions of this product
Product Code. 16671092
  • 8355.00 DKK / Each
This item is not returnable. View return policy
Description
Description
  • Built-in internal calibration at the touch of a button
  • Shock absorber function
  • Auto power ON/OFF function
  • B5 size footprint for use in narrow spaces
  • VFD Display for easy to read weighing results
  • SCF – Statistical Calculation Function
  • Standard RS-232C
  • Fast compact SHS sensor, response time only 1 second
  • Quick USB interface – no driver installation required (optional)
  • LAN – Ethernet interface with WinCT-Plus software (optional)
  • Animal weighing pan (optional)
  • Large breeze break (optional)
  • Small breeze break included on 1 mg models
  • Built-in rechargeable NiMH battery pack (optional)
Specifications
Specifications
GLP/GMP/GCP/ISO/OIML
3200 g
0.01 g
FZ-3000i-EC
11 VA to the AC adaptor (12 V, 1 A)
RS-232 C, Quick USB (option), LAN (option)
VFD
Stainless Steel
1 second
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
A&D Weighing FX/FZ Precision Balances Class II Approved > 3200 g x 0.01 g