missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin 1 Monoclonal Antibody (6C8.4-1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 437400
This item is not returnable.
View return policy
Description
The peptides used for immunization have ∽90% similarity to swine, rat, ovine, Chinese hamster sequences and 80% similar to bovine and mouse sequences.
Galectin-1 belongs to a large family of carbohydrate-binding proteins called lectins. Galectin-1can be either monomeric or homodimeric and is found in a wide variety of cells and tissue types. Galectin-1 can control cell growth, proliferation, induce apoptosis of activated T cells while it can also modulate cytokine secretion or inhibit pro-inflammatory cytokine production. Galectin-1 plays an important role in acute and chronic inflammation.Specifications
Galectin 1 | |
Monoclonal | |
0.5 mg/mL | |
LGALS1 | |
GAL1, GBP, 14 kDa laminin-binding protein, 14 kDa lectin, HBL, HLBP14, HPL, S-Lac lectin 1, beta-galactoside-binding lectin L-14-I, beta-galactoside-binding protein 14kDa, gal-1, galaptin, galectin 1, galectin-1, lactose-binding lectin 1, putative MAPK-activating protein PM12 | |
Mouse | |
Protein A | |
RUO | |
16852, 326598, 3956, 414915, 443298, 56646 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
6C8.4-1 | |
Unconjugated | |
P11116, P09382, P16045, P81184, Q49I35, P11762 | |
LGALS1 | |
Mixture of two peptides from the middle and C-terminal regions of human Galectin-1 protein | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction