missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALK1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99783JF549
This item is not returnable.
View return policy
Description
GALK1 Polyclonal specifically detects GALK1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
GALK1 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
EC 2.7.1.6, galactokinase, galactokinase 1, Galactose kinase, GALK, GK1 | |
Produced in rabbits immunized with purified, recombinant GALK1 (P51570; Met1-Leu392). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunocytochemistry, Immunofluorescence | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
2584 | |
Store at 4°C in the dark. | |
IgG |