Learn More
Thermo Scientific™ Gardnerella vaginalis Selective Medium
Isolate and identify Gardnerella vaginalis from clinical specimens with a media that contains a special blood agar base to enhance growth of fastidious organisms.
Brand: Thermo Scientific™ PB5067A
168.19 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Isolate and identify Gardnerella vaginalis from clinical specimens using Thermo Scientific™ Gardnerella vaginalis Selective Medium, a selective medium containing a special blood agar base designed to enhance the growth of fastidious organisms while maintaining osmotic balance.
Gardnerella vaginalis is found in the genitourinary tract of females and is an indicator organism for bacterial vaginosis (BV). BV is a risk factor for premature labor, chorioamnionitis, and postpartum endometritis. Women with postpartum fever and neonates with sepsis may be infected with Gardnerella vaginalis. In males, Gardnerella vaginalis has been associated with urinary tract infections, prostatitis, and urethritis. Gardnerella vaginalis is a gram-variable rod or coccobacillus. Beta hemolysis is observed on human or rabbit blood-containing media, but not sheep blood agars.Erythrocytes in the medium act as nutritional and diagnostic criterion for the production of Beta haemolysis, with Tween 80 added to improve the size of the haemolytic zones. Accompanying floara is inhibited by Gentamicin, Nalidixic acid and Amphotericin B.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
Signal red, opaque | |
Gardnerella vaginalis Selective Medium | |
Germany | |
15g ± 5% | |
10/Pk. |
2° to 12°C, in the dark | |
90mm Monoplate | |
Agar | |
Flow Wrapped - Single Layer |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6262617 | Certificate of Analysis | 09/05/2025 | PB5067A |
6260890 | Certificate of Analysis | 23/04/2025 | PB5067A |
6258067 | Certificate of Analysis | 14/04/2025 | PB5067A |
6254988 | Certificate of Analysis | 07/04/2025 | PB5067A |
6252002 | Certificate of Analysis | 31/03/2025 | PB5067A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.