Learn More
Thermo Scientific™ Gas Chromatography 3-Port SilFlow Replacement Parts
Switch your GC column in GC-MS without having to vent.
Brand:  Thermo Scientific™ 60201-401
507.03 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15207518
  • 510.00 DKK / Each
Only null left
Estimated Shipment: 31-07-2025
Description
Maintain reliability with 3-Port SilFlow replacement parts when you place your next order.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FingerTite Tool
1 Each
3-Port SilFlow Device
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Gas Chromatography 3-Port SilFlow Replacement Parts >