Learn More
Saint Gobain Life Sciences Gas Sampling Bag
Use these bags for gas sample analysis. Saint Gobain Life Sciences Gas Sampling Bag feature a cylindrical shape and an ebonite tube and tap.
Brand: Saint Gobain Life Sciences 5607073
This item is not returnable.
View return policy
Description
- Bags are individually packaged
- Available in packs of ten
- Foil thickness : 5/10mm
- Custom sizes and configurations available on request
Specifications
1 L | |
Rubber | |
Gas Sampling Bag |
Product Content Correction
Product Title
Saint Gobain Life Sciences Gas Sampling Bag > Volume: 1L
Spot an opportunity for improvement?Share a Content Correction