missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ GCKR Recombinant Protein Antigen

Recombinant Protein

Brand:  Novus Biologicals™ NBP3-17855PEP

Product Code. 18310626

  • 1664.10 DKK / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Antibody Competition
2646
100 μg
FGQTL5, GKRP, glucokinase (hexokinase 4) regulator, glucokinase (hexokinase 4) regulatory protein, Glucokinase regulator, glucokinase regulatory protein
Cancer, Cardiovascular Biology, Diabetes Research, Endocrinology, Signal Transduction
GCKR
PBS and 1M Urea, pH 7.4.
M.W. Theoretical: 30 kDa
Store at -20°C. Avoid freeze-thaw cycles.
GCKR
>80% by SDS-PAGE and Coomassie blue staining
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers