Learn More
Thermo Scientific™ GeneBLAzer™ T-Rex™ GPR119-CRE-bla CHO-K1 Cells
The GeneBLAzer™ T-Rex™ GPR119-CRE-bla CHO-K1 cells contain the human G-Protein Receptor 119 (GPR119), stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line.
Brand: Thermo Scientific™ K1486
Description
The GeneBLAzer™ T-Rex™ GPR119-CRE-bla CHO-K1 cells contain the human G-Protein Receptor 119 (GPR119), (Accession No. NP_848566) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. No. K1129) contain a beta-lactamase (bla) reporter gene under control of the cAMP response element.
The T-Rex™ GPR119-CRE-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of AR231453(1). In addition, T-Rex™ GPR119-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.
Specifications
NM_178471.1 | |
Dividing Cells | |
Fluorescence | |
GPR119 | |
Single Cell Clone Origin | |
2 x 106 Cells | |
Room Temperature | |
Dividing Cells | |
GeneBLAzer™ |
Cell-based beta lactamase reporter gene | |
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • GeneBLAzer™ T-Rex™ GPR119-CRE-bla CHO-K1 cells • GeneBLAzer™ T-Rex™ GPR119-CRE-bla CHO-K1 Assay Protocol • Certificate of Analysis The GeneBLAzer™ T-Rex™ GPR119-CRE-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
|
GPCRs | |
GPR119-CRE-bla CHO-K1 cells, GPR119-CRE-bla CHO-K1 Assay Protocol, Certificate of Analysis | |
GeneBLAzer™ | |
BLA (Beta-Lactamase) | |
GPR119 | |
CHO-K1 | |
GPCR |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1855214 | Certificate of Analysis | 19/07/2017 | K1486 |