missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Value Lab Upright Freezers
Meet routine laboratory storage requirements with upright freezers that feature digital controls and excellent temperature stability.
Brand: Thermo Scientific™ 20LFEETSV
This item is not returnable.
View return policy
Description
Meet routine laboratory storage requirements with Thermo Scientific™ Value Lab Upright Freezers. Freezers feature digital controls and excellent temperature stability.
- 20 cu. ft. upright
- Four fixed wire shelves and six door shelves
- Manual defrost
- Enameled steel interior and exterior
- CFC-free insulation and coolant
Services
13 Months Parts and Labor

Specifications
General Purpose Freezer | |
566.34 L | |
Adjustable | |
1 (1 compartment) | |
Polyurethane Foam | |
Epoxy Coated Steel | |
6 Fixed | |
Upright | |
Adjustable Leveling Feet | |
Mechanical Dial Thermostat (Inside Half Way Up Right Wall) | |
10 Years | |
Manual | |
3 in. On Sides, 3 in. At Back, 3 in. On Top | |
23.8 x 27.8 x 57.8 in. (60.3 x 70.5 x 146.7 cm) | |
5 A | |
230 V | |
50 Hz | |
Bottom-mounted | |
198 lb. | |
10 |
20 cu. ft. | |
-12°C to -20°C | |
Hinged | |
Right | |
Epoxy Coated Steel | |
4 Fixed | |
cULus | |
Temperature Display - None | |
White | |
Indoor General Storage Of Non-Critical Samples And Reagents | |
Warranty varies by country, please contact us for details | |
Interior - Yes | |
20LFEETSV | |
32.13 x 34 x 73.13 in. (81.6 x 86.4 x 185.7 cm) | |
NEMA 5-15 P | |
230 V 50 Hz | |
1150 w | |
R134a, 100% CFC Free | |
89.8 kg |
User Manual for General Purpose, Flammable Storage and Explosion Proof Freezers User Manual
Version: FEB.2016
File size: 2.5 MB
User Manual for General Purpose, Flammable Storage, and Explosion Proof Laboratory Refrigerators and Freezers
Version: FEB.2016
File size: 5.4 MB
Value Lab Upright Freezers
File size: 179.3 kB
Value Lab Upright Freezers
File size: 178.6 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Value Lab Upright Freezers > 230V/50Hz
Spot an opportunity for improvement?Share a Content Correction