Learn More
Gibertini Elettronica™ Digital Tensiometer
Click to view available options
Product Type:
Digital Tensiometer
Unit Size:
Each
Description
- Glass plates (dimensions 24 x 24 x 0.15mm)
- Glass container for liquid sample
- Manual adjustable lab jack
- Suspended system for the arrangement of glass plate
- Available with MISTEL automated jack, and WIND DCA 300 software to pilot the jack and automatically determine the surface tension and contact angle requirement
Specifications
Specifications
Product Type | Digital Tensiometer |
For Use With (Application) | Measuring liquid surface tension |
Dimensions (L x W) | Glass plate dimensions: 24 x 24 x 0.15mm |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Gibertini Elettronica™ Digital Tensiometer
Spot an opportunity for improvement?Share a Content Correction