Learn More
Gilac™ Gilactiv™ Container with Lid
Gilactiv™ Container with Lid is treated with the exclusive Sanitized™ technology, which protects the container's surface against viruses and bacteria.
667.00 DKK - 1370.00 DKK
Specifications
Type Storage Tote
Material HDPE
Color Blue
Shape Rectangular
View More Specs
Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
17918931
View Documents
Gilac™
G119690
15 L
667.00 DKK
Each
Estimated Shipment: 05-06-2025
Add to basket
17918941
View Documents
Gilac™
G119890
35 L
1140.00 DKK
Each
Estimated Shipment: 05-06-2025
Add to basket
17938771
View Documents
Gilac™
G119990
55 L
1370.00 DKK
Each
Estimated Shipment: 05-06-2025
Add to basket
Description
Description
  • Proven antibacterial and antiviral effectiveness on the container's surface (empty or full) for a broad spectrum of germs, including HCoV-229E human coronavirus.
  • This article contains a biocidal product with the active substance: Silver Phosphate Glass.
  • Tests conducted by an EU-approved French independent laboratory showed that 98.6% of the virus was wiped out after a two-hour period.
  • Choose Gilactiv™ enhanced protection.
  • Test results available on request.
  • No gaps nor overlaps and no sharp corners.
  • Rewritable identification cartridge for inventory tracking.
  • Recessed handles for better handling.
  • Smooth interior walls and rounded corners for easier cleaning.
  • Machine washable.
  • Compatible with all GILAC 4 and 6-wheel dollies.
Specifications
Specifications
Storage Tote
HDPE
Blue
Rectangular
Documents
Product Certifications
Product Content Correction
Product Title
Gilac™ Gilactiv™ Container with Lid