Learn More
Cole-Parmer™ pH Reference Half-cell
pH Reference Half-cell for research measurements.
Brand: Cole-Parmer™ 05990-60
663.96 DKK valid until 2025-04-30
Use promo code "24119" to get your promotional price.
This item is not returnable.
View return policy
Description
- Glass material.
- Half-cell.
- 6 month warranty.
- Use to build a pH electrochemical cell.
Specifications
Single | |
Glass Body | |
pH Reference Half-cell | |
Ag/AgCl Fill |
Special Offers
- Multiple Manufacturers
Product Content Correction
Product Title
Cole-Parmer™ pH Reference Half-cell > pH Reference Half-cell; Single-junction
Spot an opportunity for improvement?Share a Content Correction