Learn More
Bellingham Stanley™ Glass Centre Filling Tube With Straight Central Arm
Brand:  Bellingham Stanley™ 35-47
2260.67 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11950078
  • 2725.00 DKK / Each
Only null left
Estimated Shipment: 28-08-2025
Specifications
Center Fill Tube
Tube: Glass; End Cap: Nylon 66
Polarimetry
200 mm
Documents
One moment while we fetch your results.
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Bellingham Stanley™ Glass Centre Filling Tube With Straight Central Arm > For Use With: Polarimetry