Learn More
DWK Life Sciences Glass Ceramic Laboratory Protection Plate
The glass ceramic plates are suitable for heating of cooking glasses with the Bunsen burner.
2085.00 DKK - 2605.00 DKK
Specifications
Material | Glass Ceramics |
---|---|
For Use With (Application) | Heating glassware; Square Quadrupod for Glass Ceramic Laboratory Protection Plate |
No. per Pack | 10/Pk. |
Autoclavable | Not Autoclavable |
Product Code | Brand | Diameter (Metric) Base | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Base | Price | Quantity & Availability | ||||||
11767991
|
DWK Life Sciences
238215309 |
135 mm |
2085.00 DKK
Pack of 10 |
Estimated Shipment: 25-08-2025
Log in to see stock. |
||||||
11708041
|
DWK Life Sciences
238215703 |
155 mm |
2230.00 DKK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
12540997
|
DWK Life Sciences
238215806 |
175 mm |
2605.00 DKK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
Description
In the laboratory, solutions are often heated. This step can be carried out using different techniques, e.g. in a water or sand bath, with a heating hood or classically using a Bunsen burner. If the Bunsen burner is used as the heat source, it is recommended to use a tripod or square foot and a corresponding glass ceramic plate. The vessel to be heated is placed on the glass ceramic plate with the Bunsen burner underneath. Contamination of the glass ceramic plate can be removed with the cleaning scraper.
- Due to low thermal expansion stresses, well suited to heating glassware with a Bunsen burner
- Protects cooking glasses from direct flame
Specifications
Glass Ceramics | |
Heating glassware; Square Quadrupod for Glass Ceramic Laboratory Protection Plate | |
10/Pk. | |
Not Autoclavable |