Learn More
WTW™ Glass Overflow Measuring Flask
Replacement sample bottles and other glassware as well as useful accessories for the expansion of application of measuring systems for different parameters.WTW offers a large assortment of glass ware and other products to replace, supplement and to extend your applications.
Brand:  WTW™ 209040
326.04 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10776642
  • 342.00 DKK / Each
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Specifications
43.5 mL
BOD
Glass
Documents
One moment while we fetch your results.
Product Title
WTW™ Glass Overflow Measuring Flask > Capacity: 43.5mL