Learn More
Hamilton™ Glass Polilyte Lab Temp
Brand: Hamilton™ 242060
This item is not returnable.
View return policy
Specifications
BNC | |
Polilyte Lab Temp | |
Glass | |
Yes |
12 mm | |
120 mm | |
-10°C to +80°C |