Learn More
Gravis Glass Sample Bottles
Excellent for holding and storing small amounts of liquid samples. Gravis Glass Sample Bottles are manufactured from soda lime glass.
Brand:  Gravis E009251210
 View more versions of this product
Product Code. 15124633
  • 925.00 DKK / Pack of 210
Only null left
Estimated Shipment: 14-08-2025
Description
  • Available in a range of capacities
Specifications
Specifications
Glass
25 mL
210
210 Pk.
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Gravis Glass Sample Bottles > Capacity: 25mL