missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente™ Glass Stopcock Plug

Solid glass plug with screwthread retaining device

89.00 DKK - 123.00 DKK

Specifications

Material SBW
No. per Pack 1
View More Specs

Products 4
Product Code Brand Connectors Plug Bore Price Quantity & Availability  
Product Code Brand Connectors Plug Bore Price Quantity & Availability  
10680524
View Documents
Lenz Laborglasintrumente™
02111202
Plug NS 12.5 2.5 mm
89.00 DKK
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
 
10572793
View Documents
Lenz Laborglasintrumente™
02111402
Plug NS 14.5 2.5 mm
96.00 DKK
Each
Estimated Shipment: 13-02-2025
to see stock.
Add to basket
 
10690524
View Documents
Lenz Laborglasintrumente™
02111404
Plug NS 14.5 4.0 mm
97.00 DKK
Each
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
 
11542374
View Documents
Lenz Laborglasintrumente™
02111804
Plug NS 18.8 4.0 mm
123.00 DKK
Each
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
 
Description

Description

  • For stopcocks
  • Straight bore
  • Glass
Specifications

Specifications

SBW
1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.