Learn More
R&L Enterprises™ Glass Cutters
Provides for quick and easy cutting of glass tubing in the laboratory. R&L Enterprises™ Glass Cutters are manufactured from nickel plated zinc alloy.
Brand:  R&L Enterprises™ 3406
Product Code. 13045983
  • 215.00 DKK / Each
Only null left
Estimated Shipment: 16-09-2025
Add to basket
Description
Description
  • Accepts tubing up to 30mm in diameter
  • Versatile use in the laboratory
Specifications
Specifications
Glass Tubing Cutter
Nickel-plated Zinc
1 Ea.
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
R&L Enterprises™ Glass Cutters > Material: PTFE